Learn More
Brew Plate™ Gold Microbe Testing Kit
Click to view available options
:
Includes 5 plates and 5 tubes of semi-solid media
Description
- Detects wild yeasts, aerobic bacteria and anaerobic bacteria
- Includes 5 plates and 5 tubes of semi-solid media
Specifications
Specifications
Media Type | yeast (quad-plate), aerobic bacteria (bi-plate), and anaerobic bacteria (bi-plate) |
Product Type | Gold Microbe Testing Kit |
Includes | 5 plates and 5 tubes of semi-solid media |