Learn More
Brew Plate™Blk HLP Tubes
Click to view available options
:
filled with Hsu lactobacillus/pediococcus agar
Description
- Tubes filled with semi-solid Hsu lactobacillus/pediococcus agar
- HLP semi-solid agar detects lactobacillus and pediococcus bacteria
Specifications
Specifications
| Media Type | Hsu’s Lactobacillus/Pediococcus Agar |
| Product Type | HLP Tubes |
| Formulation | Tubes filled with semi-solid Hsu lactobacillus/pediococcus agar |
| Packaging Type | Bulk sleeve |