Learn More
Brew Plate™ Platinum Microbe Testing Kit
Click to view available options
:
Includes 5 sets of 3 plates
Description
- Detects wild yeasts, aerobic bacteria and anaerobic bacteria
- Includes five sets of 3 plates each
- Allows brewer to test up to 5 points of contamination
Specifications
Specifications
Media Type | yeast (quad-plate), aerobic bacteria (bi-plate), and anaerobic bacteria (bi-plate) |
Product Type | Platinum Microbe Testing Kit |
Includes | five sets of 3 plates each |
Quantity | 15 Plates |