Learn More
Brew Plate™ Blk Yeast Aerobic Bi-Plate
Click to view available options
:
Yeast and Aerobic Bi-Plate
Description
- Both quadrants contain Lee multi-differential agar
- Quadrant 1 contains chloramphenicol antibiotic and Quadrant 2 contains cycloheximide antifungal
Specifications
Specifications
Media Type | Chloramphenicol, Cycloheximide |
Product Type | Microbiology Media |
Formulation | Both quadrants contain Lee multi-differential agar |
Packaging Type | Bulk Sleeve |