Learn More
Brew Plate™ Blk Yeast Quad Plate
Description
- Each quadrant made from different media
- Contains Lins cupric sulfate media, Lin Wild Yeast Media; Lysine Media, and Wallterstein Nutrient Media
Specifications
Specifications
| Media Type | Lin’s Cupric Sulfate Media, Lin’s Wild Yeast Media, Lysine Media, Wallerstein Nutrient Media |
| Product Type | Microbiology Media |
| Packaging Type | Bulk Sleeve |