Learn More
Invitrogen™ BRF2 Polyclonal Antibody

Description
Immunogen sequence: VLTTTFSDEG NLREVTYSRS TGENEQVSRS QQRGLRRVRD LCRVLQLPPT FEDTAVAYYQ QAYRHSGIRA Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 87%.
Specifications
Specifications
| Antigen | BRF2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Brf2 |
| Gene Accession No. | Q9HAW0 |
| Gene Alias | 2700059M06Rik; 5730512K07Rik; AU018504; AU022602; B-related factor 2; BRF2; BRF-2; BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit; BRF2, RNA polymerase III transcription initiation factor 50 subunit; BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like; BRFU; FLJ11052; hBRFU; hTFIIIB50; PRO1470; RNA polymerase III transcription initiation factor BRFU; TFIIIB50; transcription factor IIB- related factor, TFIIIB50; transcription factor IIIB 50 kDa subunit |
| Gene Symbols | Brf2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.