Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRF2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6882R

Description
This gene encodes one of the multiple subunits of the RNA polymerase III transcription factor complex required for transcription of genes with promoter elements upstream of the initiation site. The product of this gene, a TFIIB-like factor, is directly recruited to the TATA-box of polymerase III small nuclear RNA gene promoters through its interaction with the TATA-binding protein.Specifications
BRF2 | |
Polyclonal | |
Unconjugated | |
Brf2 | |
2700059M06Rik; 5730512K07Rik; AU018504; AU022602; B-related factor 2; Brf2; BRF-2; BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit; BRF2, RNA polymerase III transcription initiation factor 50 subunit; BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like; BRFU; FLJ11052; hBRFU; hTFIIIB50; PRO1470; RNA polymerase III transcription initiation factor BRFU; TFIIIB50; transcription factor IIB- related factor, TFIIIB50; Transcription factor IIIB 50 kDa subunit | |
Rabbit | |
Protein A | |
RUO | |
55290 | |
-20°C | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9HAW0 | |
Brf2 | |
KLH conjugated synthetic peptide derived from human BRF2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction