Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brg1 Antibody (CL13779), Novus Biologicals™

Mouse Monoclonal Antibody
$342.50 - $534.50
Specifications
Antigen | Brg1 |
---|---|
Clone | CL13779 |
Dilution | Western Blot 1ug/mL, Immunocytochemistry/ Immunofluorescence 2-10 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Description
Brg1 Monoclonal antibody specifically detects Brg1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Brg1 | |
Western Blot 1ug/mL, Immunocytochemistry/ Immunofluorescence 2-10 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ATP-dependent helicase SMARCA4, BAF190, BAF190A, brahma protein-like 1, BRG1-associated factor 190A, BRG1SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 4, BRM/SWI2-related gene 1, EC 3.6.1, EC 3.6.4.-, FLJ39786, global transcription activator homologous sequence, homeotic gene regulator, hSNF2b, Mitotic growth and transcription activator, nuclear protein GRB1, Protein brahma homolog 1, Protein BRG-1, RTPS2, SNF2, SNF2B, SNF2-BETA, SNF2L4, SNF2LB, SNF2-like 4, subfamily a, member 4, sucrose nonfermenting-like 4, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI2, transcription activator BRG1 | |
This antibody was designed to target the recombinant protein Brg1, with the precise immunogen sequence being withheld for proprietary reasons. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL13779 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Cancer, Chromatin Modifiers, Neuroscience, Stem Cells | |
PBS, pH 7.2, 40% glycerol | |
6597 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title