Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRG1 Monoclonal Antibody (6D7)
GREENER_CHOICE

Catalog No. PIMA532973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532973 Supplier Invitrogen™ Supplier No. MA532973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Purity is > 95% by SDS-PAGE.

Brg1 (Brahma-related gene 1) is an ATPase subunit of SWI2/SNF2-like chromatin-remodeling complexes that enable access of regulatory and effector proteins in transcription, DNA repair and DNA replication. Although Brg1-containing complexes are not essential for general cell survival, they participate in transcriptional regulation of several hundred genes including those involved in interferon and stress response, immune cells differentiation, neurogenesis, cell cycle etc. and is absolutely necessary for mouse embryogenesis. Brg1 is also involved in cell growth arrest, senescence and tumour supression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRG1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6D7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Smarca4
Gene Accession No. P51532, Q3TKT4
Gene Alias ATP-dependent helicase SMARCA4; b2b508.1Clo; b2b692Clo; BAF190; BAF190A; brahma protein-like 1; BRG1; brg-1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; CSS4; FLJ39786; global transcription activator homologous sequence; homeotic gene regulator; HP1-BP72; hSNF2b; mitotic growth and transcription activator; MRD16; nuclear protein GRB1; protein brahma homolog 1; Protein BRG-1; RTPS2; SMARCA4; SNF2; Snf2b; SNF2beta; SNF2-beta; SNF2L4; SNF2LB; SNF2-like 4; sucrose nonfermenting-like 4; SW1/SNF; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SWI2; transcription activator BRG1
Gene Symbols Smarca4
Host Species Mouse
Immunogen Purified recombinant human SMARCA4 protein fragments expressed in E.coli.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20586, 6597
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.