Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRG1 Recombinant Rabbit Monoclonal Antibody (BLR106H)
GREENER_CHOICE

Catalog No. PIMA544403
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544403 100 μL
PIMA544404 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544403 Supplier Invitrogen™ Supplier No. MA544403
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

Brg1 (Brahma-related gene 1) is an ATPase subunit of SWI2/SNF2-like chromatin-remodeling complexes that enable access of regulatory and effector proteins in transcription, DNA repair and DNA replication. Although Brg1-containing complexes are not essential for general cell survival, they participate in transcriptional regulation of several hundred genes including those involved in interferon and stress response, immune cells differentiation, neurogenesis, cell cycle etc. and is absolutely necessary for mouse embryogenesis. Brg1 is also involved in cell growth arrest, senescence and tumour supression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRG1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR106H
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Gene Smarca4
Gene Accession No. P51532, Q3TKT4
Gene Alias ATP-dependent helicase SMARCA4; b2b508.1Clo; b2b692Clo; BAF190; BAF190A; brahma protein-like 1; BRG1; brg-1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; CSS4; FLJ39786; global transcription activator homologous sequence; homeotic gene regulator; HP1-BP72; hSNF2b; mitotic growth and transcription activator; MRD16; nuclear protein GRB1; protein brahma homolog 1; Protein BRG-1; RTPS2; SMARCA4; SNF2; Snf2b; SNF2beta; SNF2-beta; SNF2L4; SNF2LB; SNF2-like 4; sucrose nonfermenting-like 4; SW1/SNF; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SWI2; transcription activator BRG1
Gene Symbols Smarca4
Host Species Rabbit
Immunogen Peptide representing a region between residue 75 and 125 of Human transcription activator BRG1.
Purification Method purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20586, 6597
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.