Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRG1/SMARCA4 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572142
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2142 10 μL
50-157-2141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2142 Supplier Bethyl Laboratories, Inc Supplier No. IHC00466T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Dog, Horse, Rabbit, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Gray short-tailed opossum and

Brg1 (Brahma-related gene 1) is an ATPase subunit of SWI2/SNF2-like chromatin-remodeling complexes that enable access of regulatory and effector proteins in transcription, DNA repair and DNA replication. Although Brg1-containing complexes are not essential for general cell survival, they participate in transcriptional regulation of several hundred genes including those involved in interferon and stress response, immune cells differentiation, neurogenesis, cell cycle etc. and is absolutely necessary for mouse embryogenesis. Brg1 is also involved in cell growth arrest, senescence and tumour supression.

Specifications

Antigen BRG1/SMARCA4
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SMARCA4
Gene Accession No. P51532, Q3TKT4
Gene Alias ATP-dependent helicase SMARCA4; BAF190A; BRG1-associated factor 190A; BRM/SWI2-related gene 1; SNF2-like 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; brahma protein-like 1; global transcription activator homologous sequence; homeotic gene regulator; mitotic growth and transcription activator; nuclear protein GRB1; protein BRG-1; protein brahma homolog 1; sucrose nonfermenting-like 4; transcription activator BRG1
Gene Symbols SMARCA4
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20586, 6597
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.