Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRI3BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101222 Supplier Invitrogen™ Supplier No. PA5101222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BRI3B.

BRI3BP (BRI3 Binding Protein) is a Protein Coding gene located in mitochondrion. Involved in tumorigenesis and may function by stabilizing p53/TP53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRI3BP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BRI3BP
Gene Accession No. Q8BXV2, Q8WY22
Gene Alias 2410150I18Rik; AI841257; AW742481; BNAS1; BRI3 binding protein; BRI3-binding protein; Bri3bp; cervical cancer 1 proto-oncogene-binding protein KG19; cervical cancer oncogene binding protein; HCCR-1; HCCR-2; HCCRBP-1; I3-binding protein; KG19
Gene Symbols BRI3BP
Host Species Rabbit
Immunogen A synthesized peptide derived from human BRI3BP(Accession Q8WY22), corresponding to amino acid residues A13-E63.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140707, 76809
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.