Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Briakinumab Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541782
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541782 Supplier Invitrogen™ Supplier No. MA541782
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Briakinumab is a human anti-IL-12 monoclonal antibody being developed for the treatment of a number of T-cell driven autoimmune diseases. It targets and neutralize interleukin-12 and interleukin-23, two proteins associated with inflammation, such as pro-inflammatory interleukins or tumor necrosis factor- alpha. Briakinumab represents a novel approach to treating psoriasis, Multiple Sclerosis, Crohn's Disease and other autoimmune and inflammatory disorders. As of 2011 pharmaceutical development for psoriasis has been discontinued in the U. S. and Europe. Briakinumab targets and neutralizes interleukin-12 and interleukin-23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Briakinumab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias ABT-874
Host Species Human
Immunogen Human IL12B.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.