Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRICD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553951 Supplier Invitrogen™ Supplier No. PA553951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRMTLPSPHM PRPNQTILVD VARNAATITV TPPQSNHSWA VLFDGQSGCI CYRPEEHQVC FLRLMEDSDR ETLRLLVDTS KVQEAWVPSQ DTHHTQELLA VQGS Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 71%.

BRICD5 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRICD5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRICD5
Gene Accession No. Q6PL45
Gene Alias BRICD5; BRICHOS domain containing 5; BRICHOS domain-containing protein 5; BRICHOS domain-containing protein C16orf79; C16orf79
Gene Symbols BRICD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRICD5. Recombinant protein control fragment (Product #RP-91049).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283870
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.