Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Bright-Glo™ Luciferase Assay System

Catalog No. PRE2650 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL (1 Pk.)
10 x 100 mL (1 Pk.)
100 mL (1 Pk.)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-E2650 10 x 100 mL (1 Pk.)
PRE2620 100 mL (1 Pk.)
PR-E2610 10 mL (1 Pk.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-E2650 Supplier Promega Supplier No. E2650
Add to Cart
Add to Cart
Request a Quote

Includes

Bright-Glo Luciferase Assay Substrate; Bright-Glo Luciferase Assay Buffer

Bright-Glo™ Luciferase Assay System is designed for continuous-process systems. The robust, homogeneous assay chemistry gives high assay sensitivity and has a 30-minute signal half life.

  • Sensitive Reporter Assay for Continuous Processing on Robotic Systems
  • Sensitive, bright signal with 30-minute half-life
  • No need to remove culture medium or wash cells prior to adding assay reagent
  • Less sensitive to mixing conditions, sample evaporation and pipetting errors

  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  • Certain applications of this product may require licenses from others.

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Assays and biosensors that monitor cell-signaling pathways, bioluminescence resonance energy transfer (BRET) to monitor Protein-Protein interactions
Quantity 10 x 100 mL (1 Pk.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.