Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij™ 30

Catalog No. p-71941
Click to view available options
Quantity:
500 g
2.5 kg

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyoxyethylene(4)lauryl ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color Colorless to Yellow
Acid Value 2mg KOH/g max.
Physical Form Liquid
Melting Point 14.0°C
Density 0.9500g/mL
Quantity 500 g
Flash Point >149°C
Hydroxyl Value 145 to 165mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Specific Gravity 0.95
Viscosity 30 mPa.s (25°C)
Linear Formula C12H25(OCH2CH2)4OH
Water 1% max.
Fieser 01,892
Merck Index 15, 7675
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in lower alcohols, propylene glycol, insoluble in mineral oil, cottonseed oil
Formula Weight 362.54
Chemical Name or Material Brij™ 30
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
RTECSNumber MD0875000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.