Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Brij™-35, 30% Solution PROMO
Click to view available options
:
Thermo Scientific™ Brij™-35, 30% Solution
Description
Properties of Brij 35 Detergent:
- Molecular Weight: 1225 g
- Detergent Class: Non-ionic
- Aggregation Number: 40
- Micelle Molecular Weight: 49,000 g
- Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
- Cloud Point: >100°C
- Dialyzable: No
Features of this Thermo Scientific detergent solution:
- A;nonionic detergent for use in various protein methods
- Accurate—30% detergent solution in ultrapure water
- Easy-to-use—solutions are simple to dispense and dilute for use
Specifications of this Thermo Scientific Brij 35 30% solution:
- Visual: Clear, colorless viscous liquid, free of foreign material.
- Concentration: 27.0 to 30.0%
Specifications
Specifications
Product Type | Detergent |
Content And Storage | Store at room temperature |
Form | Liquid |
Product Line | Brij™, Surfact-Amps™ |
Reagent Type | Detergent Solution |
Quantity | 950 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction