Learn More
Brij∣r L23
Description
Brij™ L23 is use in Stein-Moore chromatography. Used in the schlerotherapy treatments, in those suffering from blood vessel malformations and mutations. It acts as a surfactant in Micellar electrokinetic chromatography (MEKC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Brij™ L23 is use in Stein-Moore chromatography. Used in the schlerotherapy treatments, in those suffering from blood vessel malformations and mutations. It acts as a surfactant in Micellar electrokinetic chromatography (MEKC).
Solubility
Soluble in water (40 mg/ml) at 20°C methanol, chloroform and ethanol.
Notes
Store at room temperature. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent. Used as a wetting agent, a detergent, or an emulsifier. Brij™L23 is a detergent used for measuring the concentration of carbon black in lung tissues.

Specifications
Specifications
Melting Point | ∼35°C to 40°C |
Quantity | 1000 g |
Linear Formula | CH3(CH2)10CH2(OCH2CH2)23OH |
Solubility Information | Soluble in water (40mg/ml) at 20°C methanol,chloroform and ethanol. |
Formula Weight | 1199.57 |
Chemical Name or Material | Brij™ L23 |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 500-002-6 |
RTECSNumber | JR5990000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.