Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRINP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558554 Supplier Invitrogen™ Supplier No. PA558554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLPTLLRNET GQGPVDLSDP SKRQFYIKIS DVQVFGYSLR FNADLLRSAV QQVNQSYTQG GQFYSSSSVM LLLLDIRDRI NRLAPPVAPG KPQLDLFSCM LKHRLKLTNS EIIRVNHALD LYNTEILKQS DQM Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRINP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Brinp1
Gene Accession No. O60477
Gene Alias bA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)); BMP/retinoic acid inducible neural specific 1; BMP/retinoic acid-inducible neural-specific protein (BRINP); BMP/retinoic acid-inducible neural-specific protein 1; BMP/retinoic acid-inducible neural-specific protein-1; bone morphogenetic protein/retinoic acid inducible neural-specific 1; bone morphogenic protein/retinoic acid inducible neural specific 1; bone morphogenic protein/retinoic acid inducible neural-specific 1; Brinp; Brinp1; Dbc1; Dbccr1; deleted in bladder cancer 1; deleted in bladder cancer chromosome region candidate 1; deleted in bladder cancer protein 1; Deleted in bladder cancer protein 1 homolog; Fam5a; IB3089A
Gene Symbols Brinp1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRINP1. Recombinant protein control fragment (Product #RP-92884).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1620
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.