Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRINP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550391
Description
The antibody detects endogenous levels of total DBC1 protein.
This gene is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.
Specifications
BRINP1 | |
Polyclonal | |
Unconjugated | |
Brinp1 | |
bA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)); BMP/retinoic acid inducible neural specific 1; BMP/retinoic acid-inducible neural-specific protein (BRINP); BMP/retinoic acid-inducible neural-specific protein 1; BMP/retinoic acid-inducible neural-specific protein-1; bone morphogenetic protein/retinoic acid inducible neural-specific 1; bone morphogenic protein/retinoic acid inducible neural specific 1; bone morphogenic protein/retinoic acid inducible neural-specific 1; Brinp; Brinp1; Dbc1; Dbccr1; deleted in bladder cancer 1; deleted in bladder cancer chromosome region candidate 1; deleted in bladder cancer protein 1; Deleted in bladder cancer protein 1 homolog; Fam5a; IB3089A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1620 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O60477 | |
Brinp1 | |
Full length fusion protein of Human BRINP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction