Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRINP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566437 Supplier Invitrogen™ Supplier No. PA566437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTFPECNCPDA DIQAMEDSLL QIQDSWATHN RQFEESEEFQ ALLKRLPDDR FLNSTAISQF W Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

Inhibits neuronal cell proliferation by negative regulation of the cell cycle transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRINP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRINP2
Gene Accession No. Q9C0B6
Gene Alias BMP/retinoic acid inducible neural specific 2; BMP/retinoic acid-inducible neural-specific protein 2; bone morphogenetic protein/retinoic acid inducible neural-specific 2; bone morphogenic protein/retinoic acid inducible neural-specific 2; BRINP2; DBCCR1L2; DBCCR1-like protein 2; DBCCR1-like2; FAM5B; family with sequence similarity 5, member B; KIAA1747; protein FAM5B
Gene Symbols BRINP2
Host Species Rabbit
Immunogen Recombinant Human BRINP2. Recombinant protein control fragment (Product #RP-107112).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.