Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brinzolamide 98.0+%, TCI America™
SDP

Catalog No. B4258100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4258100MG 100 mg
B425825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4258100MG Supplier TCI America Supplier No. B4258100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138890-62-7
Molecular Formula C12H21N3O5S3
Molecular Weight (g/mol) 383.496
MDL Number MFCD08067749
InChI Key HCRKCZRJWPKOAR-JTQLQIEISA-N
Synonym (R)-4-(Ethylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-Dioxide
PubChem CID 68844
ChEBI CHEBI:41212
IUPAC Name (4R)-4-(ethylamino)-2-(3-methoxypropyl)-1,1-dioxo-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide
SMILES CCNC1CN(S(=O)(=O)C2=C1C=C(S2)S(=O)(=O)N)CCCOC

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 100 mg
Formula Weight 383.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Brinzolamide
RTECSNumber XJ9095055
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.