Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRIP1 Monoclonal Antibody (pp15-IB4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA316552

Catalog No. AMSA6552


Only null left
Add to Cart

Description

Description

Suggested positive control: HeLa nuclear extract, HeLa whole cell extracts, Raji whole extracts.

LIN54 is a component of the LIN, or DREAM, complex, an essential regulator of cell cycle genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BRIP1
Monoclonal
Conc. Not Determined
ascites with 0.05% sodium azide
Q9BX63
BRIP1
A peptide containing the C-terminus of human FANCJ (within amino acids 1200-1249).
RUO
83990
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
pp15-IB4
Unconjugated
BRIP1
3110009N10Rik; 8030460J03Rik; ATP-dependent RNA helicase BRIP1; BACH 1; Bach1; BACH1 BRIP1; BRCA1 interacting protein C-terminal helicase 1; BRCA1/BRCA2-associated helicase 1; BRCA1-associated C-terminal helicase 1; BRCA1-binding helicase-like protein BACH1; BRCA1-interacting protein 1; BRCA1-interacting protein C-terminal helicase 1; BRIP 1; Brip1; FACJ; FANCJ; FANCJ protein; Fanconi anemia group J protein; Fanconi anemia group J protein homolog; FLJ90232; MGC126521; MGC126523; OF; Protein FACJ
Mouse
100 μL
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.