Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517138 Supplier Invitrogen™ Supplier No. PA517138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

BRIP1 (BRCA1 Interacting Protein C-terminal Helicase 1) is a gene situated on chromosome 17q22, covering 19 exons within its 184-kb coding region to encode a protein containing 1249 amino acids. BRIP1 is integral to the DNA repair processes, specifically through its interaction with the BRCA1 gene, and is involved in the Fanconi anemia complementation group J (FANC J). Mutations in BRIP1 have been identified, playing a role in cancer predisposition with a notable eightfold increased risk for ovarian cancer and associations with cervical cancer risk through its single nucleotide polymorphisms (SNPs). Amplifications and other mutations of BRIP1 are prevalent in a variety of cancers, and its expression involves significant correlations with copy number variations (CNV) and DNA methylation across numerous tumor types. BRIP1's expression is upregulated in certain contexts, which can contribute to cancer progression, as evidenced in the Omani population with breast cancer. These interactions validate its association with DNA damage repair, cell cycle, and metabolism across multiple cancer types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRIP1
Applications Western Blot
Classification Polyclonal
Concentration 28 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene BRIP1
Gene Accession No. Q9BX63
Gene Alias 3110009N10Rik; 8030460J03Rik; ATP-dependent RNA helicase BRIP1; BACH 1; Bach1; BACH1 BRIP1; BRCA1 interacting protein C-terminal helicase 1; BRCA1/BRCA2-associated helicase 1; BRCA1-associated C-terminal helicase 1; BRCA1-binding helicase-like protein BACH1; BRCA1-interacting protein 1; BRCA1-interacting protein C-terminal helicase 1; BRIP 1; Brip1; FACJ; FANCJ; FANCJ protein; Fanconi anemia group J protein; Fanconi anemia group J protein homolog; FLJ90232; MGC126521; MGC126523; OF; Protein FACJ
Gene Symbols BRIP1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids near the carboxy terminus of human BACH1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83990
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.