Learn More
Britton-Robinson Buffer, pH 4.10, adjusted to 0.1 M Ionic Strength, Ricca Chemical
SDP
Supplier:  Ricca Chemical Company R11544101A
 View more versions of this product
Catalog No. R11544101A
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Britton-Robinson Buffer
7732-18-5 , 7447-40-7 , 7664-38-2 , 64-19-7 , 10043-35-3 , 1310-73-2
Laboratory
4.1
Non-sterile
Colorless Liquid
98.71, 0.45, 0.3, 0.19, 0.19, 0.15
Natural Poly Bottle
1 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 24 months