Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRIX Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299257PE
Description
BRIX Polyclonal antibody specifically detects BRIX in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
BRIX | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
brix domain containing 2, Brix domain-containing protein 2, BRIXBXDC2, BRX1, biogenesis of ribosomes, homolog (S. cerevisiae), FLJ11100, ribosome biogenesis protein BRX1 homolog | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human BRIX. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
55299 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction