Learn More
Promega BRK Kinase Enzyme System, 1mg
Catalog No. PRV6276 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6276 1 mg
1 options
Catalog No. PRV6276 Supplier Promega Supplier No. V6276
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human BRK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. BRK is a member of the non-receptor tyrosine kinases (PTKs) that contains an amino terminal SH3 and SH2 domain as well as the catalytic domain.
  • Easily Screen and Profile BRK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C