Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596704 Supplier Invitrogen™ Supplier No. PA596704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRDNLLHQQM LQSEIQAMKK LRHKHILALY AVVSVGDPVY IITELMAKGS LLELLRDSDE KVLPVSELLD IAWQVAEGMC YLESQNYIHR DLAARNILVG ENTLCKVGDF GLARLIKEDV YLSHDHNIPY KWTAPEALSR GHYSTKSDVW SFGILLHEMF SRGQVPYPGM SNHEAFLRVD AGYRMPCPLE CPPSVHKLML TCWCRDPEQR PCFKALRERL SSFTSYENPT; Positive Samples: SW480, BT-474, MCF7, A-549, NIH/3T3, Mouse liver, Mouse kidney, Mouse testis, Rat kidney; Cellular Location: Cell projection, Cytoplasm, Membrane, Nucleus, ruffle.

Breast tumor kinase (Brk) is a non-receptor protein tyrosine kinase expressed in over 60% of breast carcinoma tissue samples and breast tumor cell lines. Brk is 80% identity to Sik (Src-related intestinal kinase) and exhibits the features of a novel non-receptor tyrosine kinase, including amino terminal SH3 and SH2 domains. Low levels of Brk transcripts have been identified in some human breast tumor cell lines but not in normal breast tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRK
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTK6
Gene Accession No. Q13882, Q64434
Gene Alias ALT-PTK6; breast tumor kinase; BRK; deltam5; FLJ42088; protein tyrosine kinase 6; protein-tyrosine kinase 6; protein-tyrosine kinase BRK; Ptk6; PTK6 protein tyrosine kinase 6; Sik; SRC-related intestinal kinase; tks; Tksk; tyrosine kinase identified from skin; tyrosine-protein kinase BRK
Gene Symbols PTK6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 222-451 of human PTK6 (NP_0059661).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20459, 366275, 5753
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.