Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055307
Description
BRM Polyclonal specifically detects BRM in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
BRM | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
ATP-dependent helicase SMARCA2, BAF190B, BRG1-associated factor 190B, BRMFLJ36757, EC 3.6.1, EC 3.6.4.-, global transcription activator homologous sequence, hBRMMGC74511, hSNF2a, probable global transcription activator SNF2L2, Protein brahma homolog, SNF2, SNF2/SWI2-like protein 2, SNF2A, SNF2-alpha, SNF2L2BAF190, SNF2LA, SNF2-like 2, Sth1p, subfamily a, member 2, sucrose nonfermenting 2-like protein 2, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 2, SWI2 | |
Rabbit | |
181 kDa | |
0.1 mL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, White-tufted-ear marmoset, Gray short-tailed opossum, Little brown bat, and Small-eared galago. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
P51531 | |
SMARCA2 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2 using the numbering given in entry CAA51407.1 (GeneID 6595). | |
Affinity Purified | |
RUO | |
6595 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction