Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRMS1 Mouse anti-Human, Unconjugated, Clone: 3a1.21, MilliporeSigma™

Catalog No. MABC17531MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC17531MI 100 μL
MABC17532MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC17531MI Supplier MilliporeSigma Supplier No. MABC1753100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BRMS1 Monoclonal antibody specifically detects BRMS1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRMS1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3a1.21
Conjugate Unconjugated
Gene Accession No. NP_056214
Gene Alias Breast cancer metastasis-suppressor 1
Gene Symbols BRMS1
Host Species Mouse
Immunogen GST-tagged full-length recombinant human Breast cancer metastasis-suppressor 1.
Molecular Weight of Antigen ∼35 kDa observed; 28.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 3a1.21 is a mouse monoclonal antibody that detects human Breast cancer metastasis-suppressor 1. It targets an epitope within the first 50 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.