Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRMS1L Monoclonal Antibody (OTI9A3), TrueMAB™, OriGene
SDP

Catalog No. 502543973
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3973 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3973 Supplier OriGene Supplier No. CF811037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BRMS1L Monoclonal antibody specifically detects BRMS1L in Human samples. It is validated for Western Blot

Specifications

Antigen BRMS1L
Applications Western Blot
Classification Monoclonal
Clone OTI9A3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DUSP5
Gene Accession No. Q16690
Gene Alias 0710008O11Rik; AI159718; breast cancer metastasis-suppressor 1; breast cancer metastasis-suppressor 1-like; breast cancer metastasis-suppressor 1-like protein; BRMS1; BRMS1-homolog protein p40; BRMS1L; BRMS1-like protein p40; D12Ertd407e
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 125-384 of human DUSP5 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1847
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.