Learn More
Brn3b Polyclonal Antibody, Rockland™

Goat Polyclonal Antibody
$178.35 - $649.44
Specifications
| Antigen | Brn3b |
|---|---|
| Concentration | 1.05 mg/mL |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1583
![]()
|
Rockland Immunochemicals
600101MJ0 |
100 μg |
Each for $649.44
|
|
|||||
50-199-1584
![]()
|
Rockland Immunochemicals
600101MJ0S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse Brn3b. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to rat and 91.7% to human.
POU4F2 is a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. A class IV POU-domain protein, POU4F2 is found in human retina exclusively within a subpopulation of ganglion cells where it may play a role in determining or maintaining the identities of a small subset of visual system neurons. POU4F2 is a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. A class IV POU-domain protein, POU4F2 is found in human retina exclusively within a subpopulation of ganglion cells where it may play a role in determining or maintaining the identities of a small subset of visual system neurons.[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.Specifications
| Brn3b | |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Goat | |
| Mouse, Rat | |
| G3V7L5, Q63934 | |
| 171355, 18997 | |
| Primary | |
| Antibody |
| 1.05 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| BOS_16291; Brain-3B; Brain-specific homeobox/POU domain protein 3B; brn3.2; Brn-3.2; BRN3B; brn-3b; Brn3b POU domain transcription factor; mBrn3-3R; POU class 4 homeobox 2; POU domain class 4 transcription factor 2; POU domain protein; POU domain transcription factor Brn3b-l; POU domain transcription factor Brn3b-s; POU domain, class 4, transcription factor 2; POU domain, class 4, transcription factor, related sequence 1; POU4F2; pou4f2/brn-3b; Pou4f-rs1 | |
| POU4F2 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| POU4F2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.