Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRN3B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987643
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7643 Supplier Bioss Supplier No. BS6985R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POU4F2 is a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. A class IV POU-domain protein, POU4F2 is found in human retina exclusively within a subpopulation of ganglion cells where it may play a role in determining or maintaining the identities of a small subset of visual system neurons. POU4F2 is a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. A class IV POU-domain protein, POU4F2 is found in human retina exclusively within a subpopulation of ganglion cells where it may play a role in determining or maintaining the identities of a small subset of visual system neurons.[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Specifications

Antigen BRN3B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POU4F2
Gene Accession No. Q12837
Gene Alias BOS_16291; Brain-3B; Brain-specific homeobox/POU domain protein 3B; brn3.2; Brn-3.2; BRN3B; brn-3b; Brn3b POU domain transcription factor; mBrn3-3R; POU class 4 homeobox 2; POU domain class 4 transcription factor 2; POU domain protein; POU domain transcription factor Brn3b-l; POU domain transcription factor Brn3b-s; POU domain, class 4, transcription factor 2; POU domain, class 4, transcription factor, related sequence 1; POU4F2; pou4f2/brn-3b; Pou4f-rs1
Gene Symbols POU4F2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BRN3B/POU4F2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5458
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.