Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromanil 98.0+%, TCI America™
SDP

Catalog No. T061725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T061725G 25 g
T06175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T061725G Supplier TCI America Supplier No. T061725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 488-48-2
Molecular Formula C6Br4O2
Molecular Weight (g/mol) 423.68
MDL Number MFCD00013785
InChI Key LWHDQPLUIFIFFT-UHFFFAOYSA-N
Synonym bromanil, tetrabromo-1,4-benzoquinone, bromanyl, p-bromanil, tetrabromoquinone, tetrabromo-p-benzoquinone, p-bromoanil, tetrabromo-p-quinone, tetrabromobenzoquinone, p-quinone, tetrabromo
PubChem CID 68101
IUPAC Name tetrabromocyclohexa-2,5-diene-1,4-dione
SMILES BrC1=C(Br)C(=O)C(Br)=C(Br)C1=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 423.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bromanil
RTECSNumber DK6772500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.