Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Bromo-3'-nitroacetophenone 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2227-64-7
C8H6BrNO3
244.04
MFCD00024512
GZHPNIQBPGUSSX-UHFFFAOYSA-N
2-bromo-1-3-nitrophenyl ethanone,2-bromo-3'-nitroacetophenone,3-nitrophenacyl bromide,3-nitrophenacylbromide,2-bromo-1-3-nitrophenyl ethan-1-one,a-bromo-3-nitroacetophenone,2-bromo-3-nitroacetophenone,alpha-bromo-3'-nitroacetophenone,acetophenone, 2-bromo-3'-nitro,m-nitrophenacyl bromide
75213
2-bromo-1-(3-nitrophenyl)ethanone
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr
2227-64-7
C8H6BrNO3
244.04
MFCD00024512
GZHPNIQBPGUSSX-UHFFFAOYSA-N
2-bromo-1-3-nitrophenyl ethanone,2-bromo-3'-nitroacetophenone,3-nitrophenacyl bromide,3-nitrophenacylbromide,2-bromo-1-3-nitrophenyl ethan-1-one,a-bromo-3-nitroacetophenone,2-bromo-3-nitroacetophenone,alpha-bromo-3'-nitroacetophenone,acetophenone, 2-bromo-3'-nitro,m-nitrophenacyl bromide
75213
2-bromo-1-(3-nitrophenyl)ethanone
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Melting Point 90°C to 94°C
Color Beige-Orange to Green-Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula O2NC6H4COCH2Br
Solubility Information Solubility in water: insoluble
Formula Weight 244.04
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Bromo-3'-nitroacetophenone, 97%
Show More Show Less

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

;GHS P StatementIF SWALLOWED: rinse mouth. Do NOT induce vomiting.Wear eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing.Immediatel

EINECSNumber : 218-764-0

TSCA : TSCA