Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoacetic acid, 98+%

Catalog No. AC106570010
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Pail

CAS: 79-08-3 | C2H3BrO2 | 138.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-08-3
Molecular Formula C2H3BrO2
Molecular Weight (g/mol) 138.95
MDL Number MFCD00002678
InChI Key KDPAWGWELVVRCH-UHFFFAOYSA-N
Synonym bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
PubChem CID 6227
IUPAC Name 2-bromoacetic acid
SMILES C(C(=O)O)Br

Specifications

Melting Point 49.0°C to 51.0°C
Color White to Yellow
Density 1.9300g/mL
Boiling Point 208.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrCH2CO2H
Quantity 1 kg
Beilstein 02,213
Merck Index 15,1407
Specific Gravity 1.93
Solubility Information Solubility in water: 900g/100mL (25°C). Other solubilities:g/100g(25°C): methanol-900,ether-735,benzene-308,carbon tetrachloride-168,soluble in acetone and alcohol,practically insoluble in n-heptane
Formula Weight 138.95
Percent Purity 99%
Physical Form Crystalline Mass or Crystalline Powder, Flakes, Crystals and/or Chunks
Chemical Name or Material Bromoacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
EINECSNumber 201-175-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.