Learn More
Bromoacetonitrile, 96%
Description
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
Specifications
Specifications
| Density | 1.722 |
| Boiling Point | 150°C |
| Flash Point | 81°C (178°F) |
| Refractive Index | 1.48 |
| Quantity | 50 g |
| UN Number | UN3276 |
| Beilstein | 956569 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 119.95 |
| Percent Purity | 96% |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331-H314 |
| Hazard Statement | GHS H Statement H301-H311-H331-H227-H315-H319-H335 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 209-672-1 |
| RTECSNumber | AL7970000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.