Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromobenzene, 99%

Catalog No. AAA114070C
Click to view available options
Quantity:
10,000 g
2500 g
500 g

CAS: 108-86-1 | C6H5Br | 157.01 g/mol

Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.

Solubility
Miscible with diethyl ether, alcohol, carbon tetrachloride, chloroform and benzene.

Notes
Store in cool place. Avoid inhalation of vapor or mist. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-86-1
Molecular Formula C6H5Br
Molecular Weight (g/mol) 157.01
MDL Number MFCD00000055
InChI Key QARVLSVVCXYDNA-UHFFFAOYSA-N
Synonym benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47
PubChem CID 7961
ChEBI CHEBI:3179
IUPAC Name bromobenzene
SMILES BrC1=CC=CC=C1

Specifications

Melting Point -31°C
Density 1.492
Boiling Point 155°C to 156°C
Flash Point 51°C (123°F)
Odor Aromatic
Assay Percent Range 99%
Refractive Index 1.559
Quantity 10,000 g
UN Number UN2514
Beilstein 1236661
Merck Index 14,1406
Solubility Information Miscible with diethyl ether,alcohol,carbon tetrachloride,chloroform and benzene.
Formula Weight 157.02
Percent Purity 99%
Chemical Name or Material Bromobenzene
Show More Show Less
Hazard Category H226-H315-H320
Hazard Statement GHS H Statement
H226-H315
Flammable liquid and vapor.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BROMOBENZENE
EINECSNumber 203-623-8
RTECSNumber CY9000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.