Learn More
Description
Please refer to this protocol for further information on how to use this product.
This product is part of an early adopter program .
BromoCatch™ is a trademark of Bio-Techne Corp.
Specifications
Specifications
| Chemical Name or Material | (2R)-2-((6S)-4-(4-Acrylamidophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-N-(2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)butanamide |
| Quantity | 50 μg |
| Product Line | Tocris |
| Target | BromoCatch Platform |
| Molecular Formula | C33H40N6O5S |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
