Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bromocresol Green, 0.04% Aqueous Solution

Catalog No. AC612745000
Click to view available options
Quantity:
500 mL

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-32-5
Molecular Formula C21H13Br4NaO5S
MDL Number MFCD00148898
Synonym 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt, 3', 3'', 5'

Specifications

Concentration 0.04%
Merck Index 15, 1393
Formula Weight 720
Grade Extra Pure
Packaging EcoSafPak™
Visual Transition Interval Pass Test
pH 4.3 to 4.9
Quantity 500 mL
Chemical Name or Material Bromocresol Green

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.