Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, 0.1% in Isopropanol, Reagents

Catalog No. 01335839
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
01-335-839 1 L
1 options

Catalog No. 01-335-839

Supplier: Reagents Holdings Llc CB0040001A

Only null left
Add to Cart
Add to Cart

Bromocresol Green, 0.1% in Isopropanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 76-60-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Bromocresol blue, Bromcresol Green, Bromcresol Blue
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Boiling Point 83°C
Percent Purity 0.1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.7808
Color Orange
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Bromocresol Green, 0.1% in Isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.