Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. 01336781
Click to view available options
Quantity:
1 g
5 g
10 g
100 g

Bromocresol Green, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.01
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym Bromocresol blue, Bromcresol Green, Bromcresol Blue
IUPAC Name 3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C

Specifications

Grade ACS
Packaging Glass Bottle
Specific Gravity 0.79
Color Brown to Yellow
Quantity 1 g
Physical Form Solid
Chemical Name or Material Bromocresol Green, ACS Reagent (Sultone) not water soluble
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.