Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green - Methyl Red, Alcoholic, for Alkalinity, Certified, LabChem™

Catalog No. p-5389080
Click to view available options
Quantity:
125 mL
500 mL
1 L
Packaging:
Amber Poly
White Poly

Specifications

Chemical Name or Material Bromocresol Green - Methyl Red
Melting Point -115°C
Name Note Alcoholic, for Alkalinity
Boiling Point 78°C
CAS Max % 90.36, 5, 4.5, 0.1, 0.04
CAS 67-63-0, 67-56-1, 62625-32-5, 845-10-3
Linear Formula C21H14Br4O5S·Na
UN Number UN1170
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 720.06
Grade Certified
Packaging White Poly
Specific Gravity 790kg/m3
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
Density 790kg/m3
Color Green
Quantity 125 mL
Flash Point 13°C
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.