Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromocresol Green-Methyl Red Indicator in 50% Methanol, Reagents
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Glass Bottle
Poly Bottle
Specifications
Specifications
Chemical Name or Material | Bromocresol Green-Methyl Red Indicator in 50% Methanol |
CAS | 7732-18-5, 67-56-1, 67-56-1 |
Synonym | Bromcresol Green-Methyl Red, MRBG |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 0.9 |
Quantity | 500 mL |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction