Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromocresol Green-Methyl Red TS, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Bromocresol Green-Methyl Red |
| Name Note | Test Solution |
| CAS | 64-17-5, 7732-18-5, 67-56-1, 76-60-8, 63451-28-5 |
| CAS Min % | 86.0, 11.93, 0.09, 0.06 |
| CAS Max % | 89.0, 12.42, 0.09, 0.06 |
| Grade | Indicator |
| Packaging | Natural Poly Bottle |
| Color | Red |
| Quantity | 120 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction