Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromocresol Green TS, (U.S.P. Test Solution), Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Bromocresol Green TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
Specifications
CAS Max % | 0.05%, 5%, 94.95% |
UN Number | UN1170 |
Grade | Chemical Solution |
Packaging | Poly Bottle |
Quantity | 500 mL |
Chemical Name or Material | Bromocresol Green TS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction