Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple, 0.4% w/v Aqueous, Reagents

Catalog No. 01335843
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
01-335-843 100 mL
1 options

Catalog No. 01-335-843

Supplier: Reagents Holdings Llc CB003700100A

Only null left
Add to Cart
Add to Cart

Bromocresol Purple, 0.4% w/v Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 115-40-2 , 115-40-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Bromcresol purple
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Percent Purity 0.4%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0012
Color Purple/Red
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Bromocresol Purple, 0.4% w/v Aqueous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.