Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. 01336778
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-778 5 g
01-336-779 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-778 Supplier Reagents Holdings Llc Supplier No. C61060005C2
Add to Cart
Add to Cart

Bromocresol Purple, ACS Reagent (Sultone) not water soluble, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
Synonym Bromcresol purple
IUPAC Name 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.991
Color Tan
Quantity 5 g
Physical Form Solid
Chemical Name or Material Bromocresol Purple, ACS Reagent (Sultone) not water soluble
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.