Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple sodium salt, 0.04% w/v aq. soln.

Catalog No. p-7050572
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL

CAS: 62625-30-3 | C21H15Br2NaO5S | 562.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
Molecular Weight (g/mol) 562.204
MDL Number MFCD00148896
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
Synonym Bromocresol Purple, water soluble
PubChem CID 23665568
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]

Specifications

Concentration 0.04% w/v aq. soln.
Formula Weight 562.22
Color Orange
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Bromocresol Purple sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.