Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bromocriptine mesylate
GREENER_CHOICESDP

Catalog No. 042750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-275-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-275-0 Supplier Tocris Bioscience™ Supplier No. 0427/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective D2-like agonist

Bromocriptine mesylate is a selective D3 and D2 dopamine receptor agonist (Ki values are ∼ 5 nM and ∼ 8 nM respectively). Activates both isoforms of D2 receptor (EC50 values are 0.10-0.12 nM and 4.4 nM for D2L and D2S receptors, respectively. Bromocriptine shows anti-Aβ effects that are increased in combination with Cromolyn (Cat. No. 6424) and Topiramate (Cat. No. 3620). Also lowers glucose and increases insulin sensitivity in vivo.

Chemical Identifiers

CAS 22260-51-1
Molecular Formula C33H44BrN5O8S

Specifications

Quantity 50 mg
Formula Weight 750.7
Purity >98%
Molecular Weight (g/mol) 750.706
Chemical Name or Material Bromocriptine mesylate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.